Merck
  • Rapidly equilibrating micrometer film sampler for priority pollutants in air.
Rapidly equilibrating micrometer film sampler for priority pollutants in air.
Environmental science & technology (2012-06-19)
Susan Genualdi, Tom Harner
ABSTRACT
Modified polymer-coated glass samplers (POGs), termed EVA samplers, consist of micrometer-thin layers of ethylene vinyl acetate (EVA) coated onto a glass fiber filter or aluminum foil substrate. These samplers were designed to equilibrate rapidly with priority pollutants in air, making them ideal for short-term spatial studies in ambient or indoor air. The EVA sampler was calibrated by measuring the uptake of polychlorinated biphenyls (PCBs) over 8 weeks in an indoor environment, and four different film thicknesses were monitored that ranged from 0.1 to 30 μm. The results were used to calculate the average mass transfer coefficient (50.5 m/day) and generate contour maps that provide guidance in choosing an appropriate EVA sampler for a particular study based on film thickness, deployment time, and the log K(OA) of the anlayte. A range of air pollutant classes was also added to the EVA sampler prior to deployment to assess depuration rates. These included polychlorinated biphenyls (PCBs), current-use pesticides (CUPs), perfluorinated compounds (PFCs), and polybrominated diphenyl ethers (PBDEs). On the basis of the depuration profiles, the EVA sampler was a suitable equilibrium sampler for several CUPs and PCBs; however, for the high molecular weight PCBs and PBDEs, the EVA sampler operates as a linear uptake sampler. Samplers were also evaluated for their use as a rapid screening tool for assessing concentrations of siloxanes in indoor air. The EVA sampler was used to estimate air concentrations for D4 and D5 in laboratory air to be 118 and 89 ng/m(3), respectively. Analyses were performed directly using thermal desorption gas chromatography/mass spectrometry (TDS-GC-MS). EVA samplers show promise due to their relatively low cost and ease of deployment and applicability to a wide range of priority chemicals. The ability to alter the film thickness, and hence the sorption capacity and performance of the EVA sampler, allows for a versatile sampler that can be used under varying sampling conditions and deployment times.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Pricing and availability is not currently available.