Merck
  • Copper-free click biofunctionalization of silicon nitride surfaces via strain-promoted alkyne-azide cycloaddition reactions.
Copper-free click biofunctionalization of silicon nitride surfaces via strain-promoted alkyne-azide cycloaddition reactions.
Langmuir : the ACS journal of surfaces and colloids (2012-05-31)
Radostina K Manova, Sidharam P Pujari, Carel A G M Weijers, Han Zuilhof, Teris A van Beek
ABSTRACT
Cu-free "click" chemistry is explored on silicon nitride (Si(3)N(4)) surfaces as an effective way for oriented immobilization of biomolecules. An ω-unsaturated ester was grafted onto Si(3)N(4) using UV irradiation. Hydrolysis followed by carbodiimide-mediated activation yielded surface-bound active succinimidyl and pentafluorophenyl ester groups. These reactive surfaces were employed for the attachment of bicyclononyne with an amine spacer, which subsequently enabled room temperature strain-promoted azide-alkyne cycloaddition (SPAAC). This stepwise approach was characterized by means of static water contact angle, X-ray photoelectron spectroscopy, and fluorescence microscopy. The surface-bound SPAAC reaction was studied with both a fluorine-tagged azide and an azide-linked lactose, yielding hydrophobic and bioactive surfaces for which the presence of trace amounts of Cu ions would have been problematic. Additionally, patterning of the Si(3)N(4) surface using this metal-free click reaction with a fluorescent azide is shown. These results demonstrate the ability of the SPAAC as a generic tool for anchoring complex molecules onto a surface under extremely mild, namely ambient and metal-free, conditions in a clean and relatively fast manner.
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)