- Evaluation of two enzyme immunoassays for the detection of the cocaine metabolite benzoylecgonine in 1,398 urine specimens.
Evaluation of two enzyme immunoassays for the detection of the cocaine metabolite benzoylecgonine in 1,398 urine specimens.
Benzoylecgonine (BE) is the primary urinary metabolite of cocaine. Two enzyme immunoassays were evaluated for the detection of BEin urine with a 300 ng/ml cutoff: the DRI® Cocaine Metabolite Assay and Lin-Zhi International's (LZ) Cocaine Metabolite Enzyme Immunoassay. This study involved 1,398 urine specimens from criminal justice and pain management programs. Gas chromatography/mass spectrometry (GC/MS) data were obtained for presumptive positives, and for negative urine specimens yielding responses significantly above the negative control. Approximately 46% (644) of the specimens yielded positive results by DRI, and 47% (664) were positive by LZ. One specimen screened positive with both assays but was found to have a nondetectable BE concentration by GC/MS, indicating one false positive for each assay. Twenty-one specimens yielding negative DRIresults contained BEabove 300 ng/ml, and 29 specimens yielded false negatives with the LZassay. Therefore, the overall agreement between both immunoassays and GC/MSresults was 98%. Assay sensitivity was 0.968 (DRI) and 0.958 (LZ); the selectivity for both assays was 0.999. Urine specimens containing cocaine, additional cocaine metabolites, and other drugs were also tested. No cross-reactivity was observed. Both the DRIand LZassays provide a precise, reliable method for the routine detection of BEin urine.