Merck
  • Expression of interleukin 6 and tumor necrosis factor alpha in soft tissue over ceramic and metal implant materials before uncovering: a clinical pilot study.
Expression of interleukin 6 and tumor necrosis factor alpha in soft tissue over ceramic and metal implant materials before uncovering: a clinical pilot study.
The International journal of oral & maxillofacial implants (2012-05-23)
Hans-Joachim Nickenig, Karl Andreas Schlegel, Manfred Wichmann, Stephan Eitner
ABSTRACT
To compare the effect of ceramic and metal implant materials on the expression pattern of inflammatory cytokines in peri-implant soft tissue. This split-mouth study included patients who received dental implants. In each patient, four different materials were used for the implant cover screws: the ceramic material titanium niobium oxynitride ([Ti,Nb]ON) with plasma-chemical oxidation treatment, (Ti,Nb)ON, a titanium-tantalum alloy, and tantalum. Biopsy specimens of peri-implant tissue were harvested at reentry, 4 months after the initial surgery. Immunohistochemical staining was performed for tumor necrosis factor alpha (TNF-α) and interleukin 6 (IL-6). Six patients received a total of 24 dental implants with the four different types of implant cover screws. Quantitative analysis of TNF-α; and IL-6 expression showed the lowest expression of IL-6 (1.5; interquartile range, 0 to 8.5) and TNF-α (4.8; interquartile range, 3.5 to 7.5) in tissue adjacent to the (Ti,Nb)ON layer. In comparison to the metal surfaces, the expression of both cytokines was significantly lower (P = .01 and P = .03). The lowest levels of cytokine expression were observed in tissue adjacent to the ceramic (Ti,Nb)ON coating with plasma-chemical oxidation. Based on the results of this study, this ceramic layer shows promise as a biocompatible material, and further study is warranted.
Sigma-Aldrich
Titanium nitride, <3 μm