- Methyl beta-glycosides of N-acetyl-6-O-(omega-aminoacyl)muramyl-L-alanyl-D-isoglutamines, and their conjugates with meningococcal group C polysaccharide.
Methyl beta-glycosides of N-acetyl-6-O-(omega-aminoacyl)muramyl-L-alanyl-D-isoglutamines, and their conjugates with meningococcal group C polysaccharide.
Carbohydrate research (1983-02-16)
M M Ponpipom, K M Rupprecht
PMID6839311
ABSTRACT
Spacer arms 2.1-3.7 nm (21-37 A) long were prepared, and coupled with the methyl beta-glycoside of N-acetylmuramyl-L-alanyl-D-isoglutamine benzyl ester, to give blocked 6-acylates. Deprotection was effected with palladium chloride and triethyl-silane. Chemical conjugates of MDP-meningococcal group C polysaccharide were then synthesized, in attempts to enhance the immunogenicity of the polysaccharide antigen.