Merck
  • Amino acid bound surfactants: a new synthetic family of polymeric monoliths opening up possibilities for chiral separations in capillary electrochromatography.
Amino acid bound surfactants: a new synthetic family of polymeric monoliths opening up possibilities for chiral separations in capillary electrochromatography.
Analytical chemistry (2012-05-23)
Jun He, Xiaochun Wang, Mike Morill, Shahab A Shamsi
ABSTRACT
By combining a novel chiral amino-acid surfactant containing an acryloyl amide tail, a carbamate linker, and a leucine headgroup of different chain lengths with a conventional cross-linker and a polymerization technique, a new "one-pot" synthesis for the generation of amino-acid based polymeric monolith is realized. The method promises to open up the discovery of an amino-acid based polymeric monolith for chiral separations in capillary electrochromatography (CEC). The possibility of enhanced chemoselectivity for simultaneous separation of ephedrine and pseudoephedrine containing multiple chiral centers and the potential use of this amino-acid surfactant bound column for CEC and CEC coupled to mass spectrometric detection are demonstrated.
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)