Merck
  • Highly efficient enantioselective three-component synthesis of 2-amino-4H-chromenes catalysed by chiral tertiary amine-thioureas.
Highly efficient enantioselective three-component synthesis of 2-amino-4H-chromenes catalysed by chiral tertiary amine-thioureas.
Chemical communications (Cambridge, England) (2012-05-11)
Gaosheng Yang, Chongrong Luo, Xiaolong Mu, Tingting Wang, Xin-Yuan Liu
ABSTRACT
A three-component cascade reaction of salicylaldehyde, malononitrile/cyanoacetate and nitromethane catalysed by chiral tertiary amino-thioureas was developed, which leads to the production of highly functionalized 2-amino-4H-chromenes in good yields with good to excellent enantioselectivities.
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Sigma-Aldrich
Nitromethane, ACS reagent, ≥95%
Sigma-Aldrich
Nitromethane, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Nitromethane, puriss., absolute, over molecular sieve, ≥98.5% (GC)
Sigma-Aldrich
Nitromethane, suitable for HPLC, ≥96%
Sigma-Aldrich
Malononitrile, Arxada quality, ≥99.0% (calculated, GC, KF)
Sigma-Aldrich
Malononitrile, ≥99%