Merck
  • Slow relaxation of longitudinal multispin orders in weakly and strongly coupled two-spin systems.
Slow relaxation of longitudinal multispin orders in weakly and strongly coupled two-spin systems.
Magnetic resonance in chemistry : MRC (2012-05-04)
Loyola D'Silva, Arunima Pola, Prasanta Dutta, Gary V Martinez, Peter Sprenger, Robert J Gillies, Philip W Kuchel, S Sendhil Velan
ABSTRACT
Longitudinal multispin order (LOMO) corresponds to a nonequilibrium population distribution in spin systems that exhibit scalar (J), dipolar, or quadrupolar coupling. We investigated the relaxation of longitudinal two-spin order (2-LOMO) in systems that had either weakly or strongly J-coupled spins. Our results indicated longer relaxation times for the 2-LOMO state compared with the corresponding longitudinal single-spin state (1-LOMO). Accessing nuclear spin states that have relaxation times longer than T(1), without the use of external contrast agents, is potentially useful for in vivo imaging and also for studying systems using dynamically hyperpolarized nuclear spins where longer life times are sought to increase the time available to study (bio)chemical events.
Product Description
Supelco
4-Aminobenzoic acid, analytical standard
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Aminobenzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%