Merck
Chemical and optical phototransformation of dissolved organic matter.
Water research (2012-04-17)
Steven Loiselle, Davide Vione, Claudio Minero, Valter Maurino, Antonio Tognazzi, Arduino M Dattilo, Claudio Rossi, Luca Bracchini
ABSTRACT
Dissolved organic matter represents the main reservoir of organic carbon in most aquatic ecosystems. In the present study, we determined the optical changes and the quantum yields of transient species formation for chromophoric dissolved organic matter (CDOM) samples undergoing photodegradation. The results show that the triplet states (3)CDOM* are potentially key players in CDOM photodegradation and that such transformations are strongly influenced by small differences in CDOM sources and sinks. In contrast, ·OH radicals are very unlikely to play a key role in phototransformation. These results represent an important first step in combining optical and transient species analyses to understand photodegradation processes of dissolved organic matter.
Sigma-Aldrich
Furfuryl alcohol, natural, ≥95%, FG
Sigma-Aldrich
Furfuryl alcohol, ≥97%, FG
Sigma-Aldrich
Furfuryl alcohol, 98%