Merck
  • Further electrophysiological studies on cellular effect of herbicide, bromoxynil, using characean cells.
Further electrophysiological studies on cellular effect of herbicide, bromoxynil, using characean cells.
Journal of plant research (2012-03-29)
Teruo Shimmen
ABSTRACT
In the previous paper, I reported that 3,5-dibromo-4-hydroxybenzonitrile (bromoxynil) depolarizes the plasma membrane by inhibiting the electrogenic proton pump and discussed that the inhibition is caused by cytosol acidification due to influx of protonated bromoxynil and following release of proton (Shimmen in J Plant Res 123:715-722, 2010). However, a possibility of direct inhibition of the proton pump by bromoxynil flowed into the cell could not be excluded. In the present study, the direct effect of bromoxynil on the proton pump was unequivocally excluded.
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Bromoxynil, PESTANAL®, analytical standard