Merck
  • Dispersions of nanocrystalline cellulose in aqueous polymer solutions: structure formation of colloidal rods.
Dispersions of nanocrystalline cellulose in aqueous polymer solutions: structure formation of colloidal rods.
Langmuir : the ACS journal of surfaces and colloids (2012-03-28)
Yaman Boluk, Liyan Zhao, Vanessa Incani
ABSTRACT
The steady-state shear and linear viscoelastic deformations of semidilute suspensions of rod-shaped nanocrystalline cellulose (NCC) particles in 1.0% hydroxyethyl cellulose and carboxymethyl cellulose solutions were investigated. Addition of NCC at the onset of semidilute suspension concentration significantly altered the rheological and linear viscoelastic properties of semidilute polymer solutions. The low-shear viscosity values of polymers solutions were increased 20-490 times (depending on polymer molecular weight and functional groups) by the presence of NCC. NCC suspensions in polymer solutions exhibited yield stresses up to 7.12 Pa. Viscoelasticity measurements also showed that NCC suspended polymer solutions had higher linear elastic moduli than the loss moduli. All of those results revealed the gel formation of NCC particles and presence of internal structures. The formation of a weak gel structure was due to the nonadsorbing macromolecules which caused the depletion-induced interaction among NCC particles. A simple interaction energy model was used to show successfully the flocculation of NCC particles in the presence of nonadsorbing polymers. The model is based on the incorporation of the depletion interaction term between two parallel plates into the DLVO theory for cubic prismatic rod shaped NCC particles.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~1,300,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mw ~380,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~90,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~720,000
Sigma-Aldrich
Hydroxyethyl-cellulose
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)