Merck
  • A single-blinded comparative study between the use of glycolic acid 70% peel and the use of topical nanosome vitamin C iontophoresis in the treatment of melasma.
A single-blinded comparative study between the use of glycolic acid 70% peel and the use of topical nanosome vitamin C iontophoresis in the treatment of melasma.
Journal of cosmetic dermatology (2012-03-01)
Rehab Mohamed Sobhi, Ahmed Mohamed Sobhi
ABSTRACT
Melasma is a common pigmentary disorder. Despite the availability of a wide range of skin-lightening treatments, melasma of skin remains a therapeutic challenge. The aim of this study was to evaluate the efficacy and safety of nanosome vitamin C iontophoresis and to compare the therapeutic effects of nanosome vitamin C iontophoresis vs. glycolic acid peel 70% in the treatment of melasma in Egyptian women. This study included 14 patients of melasma with skin type IV-V taken for a right-left comparison study of six sessions. Glycolic acid 70% peel was applied on the right side, whereas nanosome vitamin C was applied by iontophoresis on the other side. The results are evaluated using the melasma area and severity index score and with photographs at baseline and after six sessions. Also the photographs were evaluated by two single-blinded physicians before and after sessions. Both sides were improved, but the side treated with nanosome vitamin C showed better results. Side effects were few and transient. We concluded that nanosome vitamin C is a new, safe and effective, easy and painless method in the treatment of melasma.
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)