Merck
  • Lack of androgenicity and estrogenicity of the three monomers used in Eastman's Tritan™ copolyesters.
Lack of androgenicity and estrogenicity of the three monomers used in Eastman's Tritan™ copolyesters.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-02-22)
Thomas G Osimitz, Melanie L Eldridge, Eddie Sloter, William Welsh, Ni Ai, Gary S Sayler, FuMin Menn, Colleen Toole
ABSTRACT
Eastman Tritan™ copolyester, a novel plastic from Eastman is manufactured utilizing three monomers, di-methylterephthalate (DMT), 1,4-cyclohexanedimethanol (CHDM), and 2,2,4,4-tetramethyl-1,3-cyclobutanediol (TMCD) in various ratios. As with most any polymer, the monomers along with the high molecular weight oligomers, whose toxicity is most commonly represented by the monomers, make up the predominate amount of free chemicals available for leaching into the environment and/or foods. In light of the high level of public concern about the presence of endocrine (primarily estrogenic) activity ascribed to certain plastics and chemicals in the environment, Tritan's™ monomers were evaluated using QSAR for binding to the androgen receptor and estrogen receptors (alpha and beta) as well as a battery of in vitro and in vivo techniques to determine their potential androgenicity or estrogenicity. The findings were universally negative. When these data are coupled with other in vivo data developed to assess systemic toxicity and developmental and reproductive toxicity, the data clearly indicate that these monomers do not pose an androgenic or estrogenic risk to humans. Additional data presented also support such a conclusion for terephthalic acid (TPA). TPA is also a common polyester monomer and is the main mammalian metabolite formed from DMT.
Product Description
Supelco
Dimethyl terephthalate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%
Sigma-Aldrich
1,4-Cyclohexanedimethanol, mixture of cis and trans, 99%
Sigma-Aldrich
2,2,4,4-Tetramethyl-1,3-cyclobutanediol, mixture of isomers, 99%