- A manganese oxide with phenol groups as a promising structural model for water oxidizing complex in Photosystem II: a 'golden fish'.
A manganese oxide with phenol groups as a promising structural model for water oxidizing complex in Photosystem II: a 'golden fish'.
Dalton transactions (Cambridge, England : 2003) (2012-02-14)
Mohammad Mahdi Najafpour, Mahmoud Amouzadeh Tabrizi, Behzad Haghighi, Govindjee
PMID22327865
ABSTRACT
We describe here the ability of manganese oxide monosheets to aggregate to form layered structures with 4-aminophenol molecules. These aggregated monosheets could be considered as the first step to synthesize a self-assembled layered hybrid of phenol-manganese ions with phenol and manganese(III) and (IV) as exists in the water oxidizing complex of Photosystem II.
Product Description
Supelco
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material