Merck
  • Biochemical characterization of l-arabitol 2-dehydrogenase from Pantoea ananatis.
Biochemical characterization of l-arabitol 2-dehydrogenase from Pantoea ananatis.
Journal of bioscience and bioengineering (2012-02-07)
Yoshikiyo Sakakibara, Kyoko Torigoe
ABSTRACT
Two oxidoreductases, XDH and LAD, were found in the same operon that was involved in sugar metabolism in Pantoea ananatis. LAD, whose endogenous substrate was unknown, was recombinantly prepared and biochemically analyzed. Consequently, LAD was identified as l-arabitol 2-dehydrogenase and its substrate specificity was complementary to that of XDH.
Product Description
L-Arabinitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-(−)-Arabitol, ≥98% (GC)
Sigma-Aldrich
D-(+)-Arabitol, ≥99% (GC)