Merck
  • Removal of sudan dyes from water with C18-functional ultrafine magnetic silica nanoparticles.
Removal of sudan dyes from water with C18-functional ultrafine magnetic silica nanoparticles.
Talanta (2012-01-31)
Chunzhu Jiang, Ying Sun, Xi Yu, Lei Zhang, Xiumin Sun, Yan Gao, Hanqi Zhang, Daqian Song
ABSTRACT
In this study, the new C(18)-functionalized ultrafine magnetic silica nanoparticles (C(18)-UMS NPs) were successfully synthesized and applied for extraction of sudan dyes in water samples based on the magnetic solid-phase extraction (MSPE). The extraction and concentration were carried out in one step by blending C(18)-UMS NPs and water samples. The sudan dyes adsorbed C(18)-UMS NPs were isolated from the matrix easily with an external magnetic field. After desorption the quantitation of sudan dyes was done by ultra fast liquid chromatography (UFLC). Satisfactory extraction recovery can be obtained with only 50 mg C(18)-UMS NPs. The effects of experimental parameters, including the amount of the nanoparticles, extraction time, pH value, desorption solvent, volume of desorption solvent and desorption time were investigated. The limits of detection for sudan I, II, III and IV were 0.066, 0.070, 0.12 and 0.12 ng mL(-1), respectively. Recoveries obtained by analyzing the six spiked water samples were between 68% and 103%.
Sigma-Aldrich
Sudan III, certified by the Biological Stain Commission, BioXtra
Sigma-Aldrich
Sudan III, Technical grade
Supelco
Sudan III, analytical standard
Supelco
Sudan IV, analytical standard
Sigma-Aldrich
Sudan I, Dye content ≥95 %
Sigma-Aldrich
Sudan IV, Certified Biological Stain Commission,Dye content ≥80 %
Sigma-Aldrich
Sudan II, Dye content 90 %
Sigma-Aldrich
Sudan IV, certified by the Biological Stain Commission, BioXtra
Supelco
Sudan I, analytical standard
Supelco
Sudan II, analytical standard