Merck
  • Vaginal distribution and retention of a multiparticulate drug delivery system, assessed by gamma scintigraphy and magnetic resonance imaging.
Vaginal distribution and retention of a multiparticulate drug delivery system, assessed by gamma scintigraphy and magnetic resonance imaging.
International journal of pharmaceutics (2012-01-24)
Samata Mehta, Hans Verstraelen, Kathelijne Peremans, Geert Villeirs, Simon Vermeire, Filip De Vos, Els Mehuys, Jean Paul Remon, Chris Vervaet
ABSTRACT
For any new vaginal dosage form, the distribution and retention in the vagina has to be assessed by in vivo evaluation. We evaluated the vaginal distribution and retention of starch-based pellets in sheep as live animal model by gamma scintigraphy (using Indium-111 DTPA as radiolabel) and in women via magnetic resonance imaging (MRI, using a gadolinium chelate as contrast agent). A conventional cream formulation was used as reference in both studies. Cream and pellets were administered to sheep (n=6) in a two period-two treatment study and to healthy female volunteers (n=6) via a randomized crossover trial. Pellets (filled into hard gelatin capsule) and cetomacrogol cream, both labeled with Indium-111 DTPA (for gamma scintigraphy) or with gadolinium chelate (for MRI) were evaluated for their intravaginal distribution and retention over a 24h period. Spreading in the vagina was assessed based on the part of the vagina covered with formulation (expressed in relation to the total vaginal length). Vaginal retention of the formulation was quantified based on the radioactivity remaining in the vaginal area (sheep study), or qualitatively evaluated (women study). Both trials indicated a rapid distribution of the cream within the vagina as complete coverage of the vaginal mucosa was seen 1h after dose administration. Clearance of the cream was rapid: about 10% activity remained in the vaginal area of the sheep 12h post-administration, while after 8h only a thin layer of cream was detected on the vaginal mucosa of women. After disintegration of the hard gelatin capsule, the pellet formulation gradually distributed over the entire vaginal mucosa. Residence time of the pellets in the vagina was longer compared to the semi-solid formulation: after 24h 23 ± 7% radioactivity was detected in the vaginal area of the sheep, while in women the pellet formulation was still detected throughout the vagina. A multi-particulate system containing starch-based pellets was identified as a promising novel vaginal drug delivery system, resulting in complete coverage of the vaginal mucosa and long retention time.
Sigma-Aldrich
Brij® C10, average Mn ~683
Sigma-Aldrich
SP Brij® C2 MBAL-SO-(SG), average Mn ~330
Sigma-Aldrich
Brij® 58, average Mn ~1124
Sigma-Aldrich
ECO BRIJ® C10