Merck
  • The effect of β-carotene supplementation on the pharmacokinetics of nelfinavir and its active metabolite M8 in HIV-1-infected patients.
The effect of β-carotene supplementation on the pharmacokinetics of nelfinavir and its active metabolite M8 in HIV-1-infected patients.
Molecules (Basel, Switzerland) (2012-01-14)
Nancy L Sheehan, Rolf P G van Heeswijk, Brian C Foster, Humayoun Akhtar, Neera Singhal, Isabelle Seguin, Lina DelBalso, Marc Bourbeau, Bobby M Chauhan, Mohammed-Rachid Boulassel, David M Burger, Richard G Lalonde, Donald William Cameron
ABSTRACT
β-Carotene supplements are often taken by individuals living with HIV-1. Contradictory results from in vitro studies suggest that β-carotene may inhibit or induce cytochrome P450 enzymes and transporters. The study objective was to investigate the effect of β-carotene on the steady-state pharmacokinetics of nelfinavir and its active metabolite M8 in HIV-1 infected individuals. Twelve hour nelfinavir pharmacokinetic analysis was conducted at baseline and after 28 days of β-carotene supplementation (25,000 IU twice daily). Nelfinavir and M8 concentrations were measured with validated assays. Non-compartmental methods were used to calculate the pharmacokinetic parameters. Geometric mean ratios comparing day 28 to day 1 area under the plasma concentration-time curve (AUC(0-12 h)), maximum (C(max)) and minimum (C(min)) concentrations of nelfinavir and M8 are presented with 90% confidence intervals. Eleven subjects completed the study and were included in the analysis. There were no significant differences in nelfinavir AUC(0-12 h) and C(min) (-10%, +4%) after β-carotene supplementation. The M8 C(min) was increased by 31% while the M8 AUC(0-12 h) and C(max) were unchanged. During the 28 day period, mean CD4+ % and CD4+:CD8+ ratio increased significantly (p < 0.01). β-carotene supplementation increased serum carotene levels but did not cause any clinically significant difference in the nelfinavir and M8 exposure.
Sigma-Aldrich
Nelfinavir mesylate hydrate, ≥98% (HPLC)