Merck
  • Simple preparative gas chromatographic method for isolation of menthol and menthone from peppermint oil, with quantitative GC-MS and (1) H NMR assay.
Simple preparative gas chromatographic method for isolation of menthol and menthone from peppermint oil, with quantitative GC-MS and (1) H NMR assay.
Journal of separation science (2012-01-04)
Hae Eun Park, Seung-Ok Yang, Sun-Hee Hyun, Shin Jung Park, Hyung-Kyoon Choi, Philip J Marriott
ABSTRACT
The quantitative performance of a simple home-built preparative gas chromatography (prep-GC) arrangement was tested, incorporating a micro-fluidic Deans switch, with collection of the target compound in a deactivated uncoated capillary tube. Repeat injections of a standard solution and peppermint sample were made into the prep-GC instrument. Individual compounds were eluted from the trapping capillary, and made up to constant volume. Chloronaphthalene internal standard was added in some cases. Recovered samples were quantitatively assayed by using GC-MS. Calibration linearity of GC-MS for menthol standard area response against number of injections (2-20 repeat injections) was excellent, giving R(2) of 0.996. For peppermint, menthol correlation over 2-20 repeated injections was 0.998 for menthol area ratio (versus IS) data. Menthone calibration for peppermint gave an R(2) of 0.972. (1) H NMR spectroscopy was conducted on both menthol and menthone. Good correspondence with reference spectra was obtained. About 80 μg of isolated menthol and menthone solute was collected over a sequence of 80 repeat injections from the peppermint sample, as assayed by 600 MHz (1) H NMR analysis (∼100% recovery for menthol from peppermint). A procedure is proposed for prediction of number of injections required to acquire sufficient material for NMR detection.
Sigma-Aldrich
Peppermint oil, natural, from Mentha piperita L.
Sigma-Aldrich
Peppermint oil, purified by triple-distillation, FCC, FG
Sigma-Aldrich
Peppermint oil, natural, FCC, FG
Sigma-Aldrich
Peppermint oil, terpeneless, FCC