Merck
  • Oxidative dearomatization/intramolecular Diels-Alder cycloaddition cascade for the syntheses of (±)-atisine and (±)-isoazitine.
Oxidative dearomatization/intramolecular Diels-Alder cycloaddition cascade for the syntheses of (±)-atisine and (±)-isoazitine.
Organic & biomolecular chemistry (2011-12-24)
Xiao-Yu Liu, Hang Cheng, Xiao-Huan Li, Qiao-Hong Chen, Liang Xu, Feng-Peng Wang
ABSTRACT
A convergent and efficient formal synthesis of (±)-atisine has been accomplished. The synthetic strategy is to efficiently construct the bicyclo[2.2.2]octane ring moiety by an oxidative dearomatization/intramolecular Diels-Alder cycloaddition cascade. The first total synthesis of another atisine-type C(20)-diterpenoid alkaloid, (±)-isoazitine, has also been achieved employing the same strategy.
Product Description
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Octane, reagent grade, 98%
Sigma-Aldrich
Octane, anhydrous, ≥99%