Merck
  • Improving stability and activity of cross-linked enzyme aggregates based on polyethylenimine in hydrolysis of fish oil for enrichment of polyunsaturated fatty acids.
Improving stability and activity of cross-linked enzyme aggregates based on polyethylenimine in hydrolysis of fish oil for enrichment of polyunsaturated fatty acids.
Applied biochemistry and biotechnology (2011-12-15)
Jinyong Yan, Xiaohua Gui, Guilong Wang, Yunjun Yan
ABSTRACT
Cross-linking of enzyme aggregates from recombinant Geotrichum sp. lipase based on polyethylenimine (PEI) was applied to hydrolyze fish oil for enrichment of polyunsaturated fatty acids successfully. Through acetone precipitation and cross-linking of physical aggregates using glutaraldehyde in the presence of PEI, firmly cross-linked enzyme aggregates (PEI-CLEAs) were prepared. They could maintain more than 65% of relative hydrolysis degree after incubation in the range of 50-55 °C for 4 h and maintain more than 85% of relative hydrolysis degree after being treated by acetone, tert-butyl alcohol and octane for 4 h. PEI-CLEAs increased hydrolysis degree to 42% from 12% by free lipase. After five batch reactions, PEI-CLEAs still maintained 72% of relative hydrolysis degree. Hydrolysis of fish oil by PEI-CLEAs produced glycerides containing concentrated EPA and DHA in good yield. PEI-CLEAs had advantages over general CLEAs and free lipase in initial reaction rate, hydrolysis degree, thermostability, organic solvent tolerance and reusability.
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Octane, reagent grade, 98%
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%