Merck
  • Effects of Na₂CO₃ stress on photosynthesis and antioxidative enzymes in endophyte infected and non-infected rice.
Effects of Na₂CO₃ stress on photosynthesis and antioxidative enzymes in endophyte infected and non-infected rice.
Ecotoxicology and environmental safety (2011-12-06)
Ning Bu, Xuemei Li, Yueying Li, Chunyan Ma, Lianju Ma, Chi Zhang
ABSTRACT
Endophyte infected and uninfected seedlings of rice (Oryza sativa L.) were subjected to five different levels of Na(2)CO(3) for 2 weeks. Under both Na(2)CO(3) stress as well as no stress, endophyte-infected plants were higher for above-ground dry weight and shoot length, but lower for root length compared to non-infected controls, and there was no significant difference for below-ground dry weight. Chlorophylls and carotenoid contents, net photosynetic rate, transpiration rate, catalase and peroxidase activities increased, but malondialdehyde content declined in the infected plants compared to non-infected controls under Na(2)CO(3) stress. Compared to non-infected controls, water use efficiency, stomatal conductance, chlorophyll fluorescence parameters (F(v)/F(m), F(v)/F(o)) and superoxide dismutase activity in infected plants increased under high concentrations of Na(2)CO(3) while there was no significant difference under low concentrations of Na(2)CO(3). Endophyte infection was concluded to be beneficial to the growth and antioxidative mechanisms in Oryza sativa under Na(2)CO(3) stress.
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7