Merck
  • [The mechanical properties and moisture permeability of eudragit L100/S100 free films affected by plasticizers and membrane materials ratio].
[The mechanical properties and moisture permeability of eudragit L100/S100 free films affected by plasticizers and membrane materials ratio].
Yao xue xue bao = Acta pharmaceutica Sinica (2011-11-30)
Guo-song Zhang, Chuan-hua Feng, Wei Jiang, Peng-yi Hu, Ping Deng, Yao Zhang, Xiao-jian Luo
ABSTRACT
The free membrane of Eudragit L100/S100 which is pH-sensitive, colon-specific was prepared by plane casting films. The film humidity, species and amount of plasticizers, the ratio of membrane material was investigated. The rate of membrane permeability and mechanical properties were used as indicators of orthogonal experiment, and its related properties were studied. The results show that the mechanical properties of the membrane and phragmoid capacity are the best when 30% TEC was used as plasticizer; the ratio of membrane material have little effect on the rate of membrane permeability and mechanical properties. By adjusting the species and amount of plasticizers, the ratio of Eudragit L100/S100, the free membrane which is colon-specific can be obtained.
Product Description
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Glyceryl triacetate, ≥99.0% (GC)
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Glyceryl triacetate, ≥99%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Sigma-Aldrich
Triacetin, 99%
Sigma-Aldrich
Triethyl citrate, ≥99%, FCC, FG
Sigma-Aldrich
Triacetin, 99%, FCC, FG
Sigma-Aldrich
Triethyl citrate, ≥98.0% (GC)
Supelco
Triethyl citrate, Pharmaceutical Secondary Standard; Certified Reference Material