Merck
  • Micro-Raman spectroscopic analysis of the degree of conversion of composite resins containing different initiators cured by polywave or monowave LED units.
Micro-Raman spectroscopic analysis of the degree of conversion of composite resins containing different initiators cured by polywave or monowave LED units.
Journal of dentistry (2011-11-19)
Vesna Miletic, Ario Santini
ABSTRACT
To determine the degree of conversion (DC) over 48 h post-curing of resin mixtures containing trimethylbenzoyl-diphenylphosphine oxide (TPO) initiator cured by a polywave or a monowave LED light-curing unit (LCU). In resin mixtures based on equal weight percent (wt%) of BisGMA and TEGDMA the following initiators were added: 0.2 wt% camphorquinone (CQ)+0.8 wt% ethyl-4-dimethylaminobenzoate (EDMAB) (Group 1); 1 wt% TPO (Group 2) and 0.1 wt% CQ+0.4 wt% EDMAB+0.5 wt% TPO (Group 3). Half of the samples in each group (n=5) were cured using a polywave (bluephase(Ā®) G2, Ivoclar Vivadent) or a monowave LED LCU (bluephase(Ā®), Ivoclar Vivadent). The DC was measured using micro-Raman spectroscopy within 5 min and then 1, 3, 6, 24 and 48 h post-irradiation. The data were analysed using general linear model and two-way ANOVA for the factors 'time', 'material', 'surface' and 'LCU' at Ī±=0.05. The initial DC values obtained upon light curing remained similar over a 48 h period. bluephase(Ā®) G2 produced the highest DC in Group 2 followed by Group 3, and Group 1. bluephase(Ā®) resulted in the highest DC in Group 1, followed by Group 2 and Group 3 (p<0.05). Unfilled resin materials containing both TPO- and CQ-amine initiators are effectively cured using bluephase(Ā®) G2. Resin mixture with the same wt% of initiators is better cured when TPO is the only initiator, compared to CQ-amine only or combined TPO and CQ-amine system. After initial light cure, no additional conversion of uncured monomers was detected in an unfilled resin material over 48 h at 37Ā°C.
Sigma-Aldrich
(1S)-(+)-Camphorquinone, 99%