Merck
  • Clinical feasibility of monitoring m-THPC mediated photodynamic therapy by means of fluorescence differential path-length spectroscopy.
Clinical feasibility of monitoring m-THPC mediated photodynamic therapy by means of fluorescence differential path-length spectroscopy.
Journal of biophotonics (2011-08-24)
Baris Karakullukcu, Stephen Chad Kanick, Jan Bonne Aans, Henricus Jcm Sterenborg, I Bing Tan, Arjen Amelink, Dominic J Robinson
ABSTRACT
The objective quantitative monitoring of light, oxygen, and photosensitizer is challenging in clinical photodynamic therapy settings. We have previously developed fluorescence differential path-length spectroscopy (FDPS), a technique that utilizes reflectance spectroscopy to monitor microvascular oxygen saturation, blood volume fraction, and vessel diameter, and fluorescence spectroscopy to monitor photosensitizer concentration. In this paper the clinical feasibility of the technique is tested on eight healthy volunteers and on three patients undergoing PDT of oral cavity cancers. Model-based analysis of the measured spectra provide quantitative tissue parameters that are corrected for background tissue absorption, autofluorescence, and the transmission of the optical system; this method allows comparison of intra- and inter-subject parameters. The FDPS correctly estimated the absence of m-THPC in volunteers and detected photobleaching in the areas receiving treatment light in patients undergoing PDT treatment. This study demonstrates the feasibility of monitoring clinical photodynamic therapy treatments using optical spectroscopy.
Sigma-Aldrich
Tetrakis(hydroxymethyl)phosphonium chloride solution, 80% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution, technical, 70-75% in H2O (T)
Pricing and availability is not currently available.