Merck
  • Essential oil composition of the fruits of Periploca laevigata Aiton subsp. angustifolia (Labill.) Markgraf (Apocynaceae - Periplocoideae).
Essential oil composition of the fruits of Periploca laevigata Aiton subsp. angustifolia (Labill.) Markgraf (Apocynaceae - Periplocoideae).
Natural product research (2011-08-24)
Pietro Zito, Maurizio Sajeva, Maurizio Bruno, Antonella Maggio, Sergio Rosselli, Felice Senatore, Carmen Formisano
ABSTRACT
The essential oil of the fruits of Periploca laevigata Aiton subsp. angustifolia (Labill.) Markgraf (Apocynaceae) from Lampedusa Island was obtained by hydrodistillation and its composition was analysed. The analyses allowed the identification and quantification of 64 volatile compounds belonging to different classes. The most abundant compounds were nonacosane, heptacosane, hentriacontane and δ-cadinene. Among the volatile compounds identified in the fruits of P. laevigata subsp. angustifolia, 31 are present in other taxa of Apocynaceae, 19 have antimicrobial activity and four are pheromones for the butterfly Danaus chrysippus. The possible ecological role of the volatile compounds found is briefly discussed.
Product Description
Supelco
Hentriacontane, analytical standard
Sigma-Aldrich
Nonacosane, 99%
Supelco
Nonacosane, analytical standard