Merck
Theoretical study of magnesium fluoride in aqueous solution.
The journal of physical chemistry. B (2011-08-19)
Naoto Shibata, Hirofumi Sato, Shigeyoshi Sakaki, Yuji Sugita
ABSTRACT
A series of magnesium fluorides (MgF(n)(2-n)), multiply charged anions, in the gas phase and in aqueous solution were theoretically studied with a hybrid approach of quantum chemistry and statistical mechanics, called RISM-SCF-SEDD theory. In the gas phase, MgF(3)(-) is the most stable species among the complexes (n = 1-6). In contrast, due to compensation between the intramolecular energy and solvation free energy, the stabilities of a number of complexes with different n are comparable in aqueous solution. Based on accurate evaluation of free energy change, the mole fraction of MgF(4)(2-) is the highest in the range from pF = 2.0 to 3.0 of aqueous solution. This is consistent with the available PDB data of the enzymes that catalyze the phosphoryl transfer reactions. The hydration structures of magnesium fluorides obtained by RISM-SCF-SEDD theory provide insight into their structural changes from the gas phase to aqueous solution.
Sigma-Aldrich
Magnesium fluoride, random crystals, optical grade, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium fluoride, technical grade
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium fluoride, pieces, 3-6 mm, 99.9% trace metals basis (excluding Na)
Pricing and availability is not currently available.