Merck
  • The therapeutic effects of R8-liposome-BCG-CWS on BBN-induced rat urinary bladder carcinoma.
The therapeutic effects of R8-liposome-BCG-CWS on BBN-induced rat urinary bladder carcinoma.
Anticancer research (2011-07-09)
Jun Miyazaki, Hiroyuki Nishiyama, Ikuya Yano, Akihiro Nakaya, Hideyasu Kohama, Koji Kawai, Akira Joraku, Takashi Nakamura, Hideyoshi Harashima, Hideyuki Akaza
ABSTRACT
The present gold standard for bladder cancer is Mycobacterium bovis bacillus Calmette-Guérin (BCG) immunotherapy, but serious side-effects are common. We previously reported that C3H/HeN mice vaccinated with a mixture of MBT-2 cells and artificial BCG, octaarginine-modified liposomes incorporating the cell wall of BCG (R8-liposome-BCG-CW), significantly inhibited growth of R8-liposome-BCG-CW pretreated MBT-2 cells. Our aim was to determine if a non-live bacterial agent could be as efficacious as live BCG in a model of bladder cancer. We investigated the suppressive effect of liposome-incorporating cell wall skeleton (BCG-CWS) on N-butyl-N-(4-hydroxybutyl) nitrosamine (BBN)-induced urinary bladder carcinogenesis in rats. F344 rats were fed with BBN and sodium ascorbate for 8 weeks, after which all rats were confirmed to have excreted atypical epithelial cells in the urine. Rats were administered BCG-CW(1.0 mg/rat) or R8-liposome-BCG-CWS (0.1 or 1.0 mg/rat) intravesically once/week for 8 weeks from week 28 to 35 of the experimental protocol. Rats receiving R8-liposome-BCG-CWS intravesically showed significantly inhibited numbers of tumors, especially those of simple hyperplasia, in comparison with the control rats. R8-liposome-BCG-CWS administration had inhibitory effects on rat bladder carcinogenesis. These results may indicate a novel adoptive immunotherapy against bladder cancers.
Sigma-Aldrich
N-Butyl-N-(4-hydroxybutyl)nitrosamine, ISOPAC®, ≥90% (GC)