Merck
  • Molecular mechanism of uncaging CO2 from nitrophenylacetate provides general guidelines for improved ortho-nitrobenzyl cages.
Molecular mechanism of uncaging CO2 from nitrophenylacetate provides general guidelines for improved ortho-nitrobenzyl cages.
Chemphyschem : a European journal of chemical physics and physical chemistry (2011-06-10)
Jan-Michael Mewes, Karsten Neumann, Mirka-Kristin Verhoefen, Georg Wille, Josef Wachtveitl, Andreas Dreuw
ABSTRACT
Femtosecond spectroscopy and quantum chemical calculations provide detailed insights into the specificities of the uncaging mechanism of CO2 from ortho-, meta-, and para-nitrophenylacetate. The emerging general principles allow a rational design of improved ortho-nitrophenyl cages for chemical and biological applications.
Sigma-Aldrich
2-Nitrophenylacetic acid, 98%