Merck
  • Photosensitized amino acid degradation in the presence of riboflavin and its derivatives.
Photosensitized amino acid degradation in the presence of riboflavin and its derivatives.
Environmental science & technology (2011-05-20)
Christina K Remucal, Kristopher McNeill
ABSTRACT
The addition of photosensitizers to water can accelerate disinfection in sunlight-based systems by enhancing oxidation of target compounds through direct reaction with the excited sensitizer or through production of another oxidant, such as singlet oxygen (¹O₂). The kinetics of the oxidation of selected amino acids in the presence of the sensitizer riboflavin (Vitamin B2), its primary photoproduct lumichrome, and its derivative riboflavin tetraacetate (2',3',4',5'-tetraacetylriboflavin; RTA) were quantified and the mechanisms of reaction were determined during exposure to 365 ± 9 nm light. ¹O₂-mediated reactions contributed to the rapid photodegradation of the four amino acids, but its contribution was sensitizer-dependent and varied from 5.4-10.2% for tyrosine, 7.1-12.4% for tryptophan, 18.7-69.0% for methionine, and 64.7-100.2% for histidine. Riboflavin was subject to rapid photodegradation (t½ < 8 min), while the half-lives of lumichrome and RTA were 100 and 30 times longer, respectively. Lumichrome and RTA also were more efficient ¹O₂ sensitizers (quantum yield (Φ) = 0.63 and 0.66) compared to riboflavin (Φ = 0.48). Of the three flavin-based compounds, RTA shows the most promise as a sensitizer in sunlight-based disinfection systems because it absorbs both visible and UV light, is an efficient ¹O₂ sensitizer, is a strong oxidant in its triplet state, and exhibits greater photostability.
Sigma-Aldrich
Lumichrome
Sigma-Aldrich
Furfuryl alcohol, 98%
Sigma-Aldrich
Furfuryl alcohol, natural, ≥95%, FG
Sigma-Aldrich
Furfuryl alcohol, ≥97%, FG