Merck
  • Novel molecularly imprinted microsphere using a single chiral monomer and chirality-matching (S)-ketoprofen template.
Novel molecularly imprinted microsphere using a single chiral monomer and chirality-matching (S)-ketoprofen template.
Journal of chromatography. A (2011-05-11)
Jiabing Jiang, Kesheng Song, Zhi Chen, Qi Zhou, Youwen Tang, Fenglong Gu, Xiongjun Zuo, Zhiguang Xu
ABSTRACT
We designed and synthesized a cinchonine derivative to be used as a novel chiral monomer. It was employed in a dual role of functional monomer and cross-linking monomer, displaying multi-binding sites for the template (S)-ketoprofen. Monodisperse molecularly imprinted core-shell microspheres were prepared using surface imprinting method on silica gel. The results show a substantial synergistic effect in the enantioselective recognition, confirming our initial hypothesis. Computational simulation of the monomer and template pre-arrangement strongly supports our proposed chiral recognition mechanism for the imprinted microspheres.
Sigma-Aldrich
(+)-Cinchonine, 85%
Pricing and availability is not currently available.
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)
Pricing and availability is not currently available.