Merck
  • Risk of egg parasitoid attraction depends on anti-aphrodisiac titre in the large cabbage white butterfly Pieris brassicae.
Risk of egg parasitoid attraction depends on anti-aphrodisiac titre in the large cabbage white butterfly Pieris brassicae.
Journal of chemical ecology (2011-04-01)
Martinus E Huigens, Erik de Swart, Roland Mumm
ABSTRACT
Males of a variety of insects transfer an anti-aphrodisiac pheromone to females during mating that renders them less attractive to conspecific males. In cabbage white butterflies, the transfer of an anti-aphrodisiac can result in the unwanted attraction of tiny egg parasitoid wasps of the genus Trichogramma that hitch-hike with mated female butterflies to a host plant where they parasitize the freshly laid butterfly eggs. Here, we show that the anti-aphrodisiac benzyl cyanide (BC) of the large cabbage white Pieris brassicae is depleted by frequent display of the mate-refusal posture that signals a female's unreceptivity to mating. This depletion of BC is ecologically important because it results in a reduced risk of attracting the hitch-hiking egg parasitoid Trichogramma brassicae to mated female butterflies over time since mating. Our results indicate for the first time that a reduction in anti-aphrodisiac titre in mated females due to frequent adoption of the mate-refusal posture is beneficial to both mated females and males particularly when parasitoid pressure is high.
Sigma-Aldrich
Benzyl cyanide, 98%
Pricing and availability is not currently available.