Merck
  • Isotropic photo-decomposition of spherical organic polymers on rutile TiO₂(110) surfaces.
Isotropic photo-decomposition of spherical organic polymers on rutile TiO₂(110) surfaces.
Nanotechnology (2011-03-11)
Nobuyuki Ishida, Tamaki Iwasaki, Daisuke Fujita
ABSTRACT
We observed the photo-decomposition process of polystyrene latex (PSL) spheres on a rutile TiO₂(110) single crystal surface by using atomic force microscopy. During the decomposition process, both the height and width of the PSL spheres linearly decreased with the irradiation time in a similar way from the beginning, suggesting that the PSL spheres are isotropically decomposed. This indicates that the interface between the PSL spheres and the TiO₂ surface is not a dominant reaction site, as expected from normal photocatalytic reactions.
Sigma-Aldrich
Polystyrene, average Mw ~280,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~400,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000