Merck
  • Ligament reconstruction tendon interposition with mersilene augmentation.
Ligament reconstruction tendon interposition with mersilene augmentation.
Techniques in hand & upper extremity surgery (2011-03-02)
Andrew J Stein, Jennifer L Schofield, Mike Marsh, Jerry Paulo
ABSTRACT
Many surgical procedures have been described for the treatment of thumb basilar joint osteoarthritis. Augmentation of the standard ligament reconstruction tendon interposition procedure with the use of a Mersilene suture tape suspension-plasty, to recreate the stability provided by the anterior oblique ligament and increase pinch strength, will be described. Satisfaction with this procedure was evaluated through surveys completed by patients. In addition, independent physical assessments were performed to demonstrate stability, range of motion, and strength.
Sigma-Aldrich
Poly(ethylene terephthalate), granular