Merck
Adsorption of p-cresol on novel diatomite/carbon composites.
Journal of hazardous materials (2011-02-23)
H Hadjar, B Hamdi, C O Ania
ABSTRACT
Hybrid inorganic/organic adsorbents were synthesized using mixtures of diatomite and carbon charcoal as precursors, and explored for the removal of p-cresol from aqueous solution. The carbon/diatomite composites displayed a bimodal and interconnected porous structure which was partially inherited from both precursors. They display moderate surface areas (between 100 and 400 m(2)g(-1)) due to their large inorganic content (between 70 and 90 wt.%), since the diatomite is a non-porous material. Compared to activated carbons with a more developed porosity, p-cresol adsorption on the prepared carbon/diatomite composites was much faster, showing adsorption capacities similar to those of conventional adsorbents over a wide pH range. These results show a good affinity of p-cresol molecules towards the hybrid inorganic/organic composites, and demonstrate the suitability of these novel materials for the removal of aromatic (polar) molecules, despite their dominant inorganic character.
Product Description
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 535, filter aid, flux calcined