- Determination of field-incurred chlorfluazuron residues in the peach.
Determination of field-incurred chlorfluazuron residues in the peach.
Bulletin of environmental contamination and toxicology (2011-02-18)
Jeong-Heui Choi, Md Iqbal Rouf Mamun, Jong-Hyouk Park, Eun-Ho Shin, Jae-Han Shim
PMID21327612
ABSTRACT
Chlorfluazuron residues were determined in the peaches that were sprayed at dosage (a.i. 0.0167 kg 10a(-1)), using an analytical method that was validated as follows; r (2) = 0.9999, 0.02 mg kg(-1) (LOQ) and 87.8-93.6% (recovery). The residues from all samples were lower than the MRL (0.5 mg kg(-1), Korea). A maximum 0.27 mg kg(-1) of chlorfluazuron was detected in the samples applied at 6 days before harvest. The results signify that the 10% SC product would be used safely as an insecticide if it is applied two or three times onto peaches, with applications given until 6 days prior to harvest.
MATERIALS