Merck
  • Five carboxin-resistant mutants exhibited various responses to carboxin and related fungicides.
Five carboxin-resistant mutants exhibited various responses to carboxin and related fungicides.
Bioscience, biotechnology, and biochemistry (2011-01-14)
Yoko Shima, Yasuhiro Ito, Hidemi Hatabayashi, Akemi Koma, Kimiko Yabe
ABSTRACT
Five carboxin-resistant mutants from Aspergillus oryzae were characterized by the sensitivities of their mycelial growth and succinate dehydrogenase (SDH) activity to carboxin and three related fungicides. Despite a significant resistance to carboxin, exhibited by all the mutants, their patterns of sensitivity to the other fungicides was distinct. This provides clues to the molecular interaction between SDH and these fungicides.
Product Description
Supelco
Carboxine, PESTANAL®, analytical standard