Merck
  • Anticholinesterase activity of standardized extract of Illicium verum Hook. f. fruits.
Anticholinesterase activity of standardized extract of Illicium verum Hook. f. fruits.
Fitoterapia (2010-11-16)
Santanu Bhadra, Pulok K Mukherjee, N Satheesh Kumar, A Bandyopadhyay
ABSTRACT
Illicium verum is a well known spice in traditional Indian system for its therapeutic potential. The present study was aimed to evaluate the acetylcholinesterase (AChE) and butyrylcholinesterase inhibitory (BChE) activity of standardized extracts of I. verum and its oil. Present study confirmed that anethole contributed to the anticholinesterase activity of I. verum, with more specificity towards AChE. IC(50) for AChE and BChE inhibitory activity of anethole was 39.89±0.32 μg/mL and 75.35±1.47 μg/mL, whereas for the oil, 36.00±0.44 μg/mL and 70.65±0.96 μg/mL respectively. Therefore I. verum can be a good lead as anti-cholinesterase agent from natural resources.
Product Description
Supelco
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Supelco
trans-Anethole, analytical standard