Merck
  • Dicarboxy-dichlorofluorescein: a new fluorescent probe for measuring acidic intracellular pH.
Dicarboxy-dichlorofluorescein: a new fluorescent probe for measuring acidic intracellular pH.
Analytical biochemistry (1990-05-15)
M Nedergaard, S Desai, W Pulsinelli
ABSTRACT
Derivatives of fluorescein sensitive to pH are extensively utilized for the determination of intracellular pH (pHi). Available dyes have pKa values of approximately 7.0, and are not well suited for measuring acidic pHi. We examined the fluorescein derivative, 5 (and 6)-carboxy-2',7'-dichlorofluorescein (CDCF) for its potential in the microspectrofluorometric measurement of pHi during acidic conditions. CDCF showed intense fluorescence and pH sensitivity near its "effective" pKa value of 4.2, using a 495/440 nm dual excitation wave-length ratio method. Protein interactions caused fluorescence ratio deviations which were most pronounced at the extremes of pH, whereas calcium and magnesium concentrations had little effect on the fluorescent ratio intensity. Intracellular calibration performed using nigericin in the presence of high potassium eliminated the need to correct for protein interactions, and the ratio method minimized any variations due to dye concentration differences or instrument fluctuation. Intracellular retention of the dye was high, and 95% of the initial signal remained after 1 h. Fluorescence bleaching was 14.5% after 1 h of continuous excitation and cell survival was not affected by dye loading. We conclude that CDCF is an excellent intracellular pH indicator in the pH range of 4-5.
Sigma-Aldrich
5(6)-Carboxy-2′,7′-dichlorofluorescein, BioReagent, suitable for fluorescence, ≥95% (TLC)