Merck
  • [Studies on the chemical constituents of rhizoma of Coniogramme japonica].
[Studies on the chemical constituents of rhizoma of Coniogramme japonica].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-09-18)
Cheng-Wu Fang, Jia-Jia Chen, Shou-Jin Liu
ABSTRACT
To study the chemical constituents of Rhizoma of Coniogramme japonica. The Rhizome of C. japonica were extracted with ethanol and the chemical constituents were separated by chromatography technique, and their structures were determined by spectral analysis. Six compounds were isolated and identified as butyl 2-formyl-5-butoxymethyl-1 H-pyrrole-1-butanoate (1), pterolactam (2), 5-hydroxymethylfurfural (3), methyl linoleate (4), trilinolein (5), dehydrovomifoliol (6). All compounds are isolated from the genus for the first time, and the compound 1 is a new compound.
Product Description
Supelco
Methyl linoleate, analytical standard
Supelco
Methyl linolelaidate, analytical standard
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Sigma-Aldrich
Methyl linolelaidate
Supelco
trans-9,12-Octadecadienoic acid methyl ester, certified reference material, 10 mg/mL in heptane
Sigma-Aldrich
Glyceryl trilinoleate, ≥98% (TLC), liquid
Supelco
cis-9,cis-12-Octadecadienoic acid methyl ester, certified reference material, 10 mg/mL in heptane