Merck
  • One-pot AgOAc-mediated synthesis of polysubstituted pyrroles from primary amines and aldehydes: application to the total synthesis of purpurone.
One-pot AgOAc-mediated synthesis of polysubstituted pyrroles from primary amines and aldehydes: application to the total synthesis of purpurone.
Organic letters (2010-08-26)
Qingjiang Li, Aili Fan, Zhiyao Lu, Yuxin Cui, Wenhan Lin, Yanxing Jia
ABSTRACT
A simple and efficient method for the synthesis of 1,3,4-trisubstituted or 3,4-disubstituted pyrroles has been developed. The reaction represents the first time that pyrroles are synthesized directly from readily available aldehydes and amines (anilines) as starting materials. This method has been successfully applied to the rapid synthesis of purpurone.
Sigma-Aldrich
Silver acetate, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silver acetate, ReagentPlus®, 99%
Pricing and availability is not currently available.