Merck
  • Enhanced photodegradation of dyes on titania-based photocatalysts by adding commercial GeO2 in aqueous suspension.
Enhanced photodegradation of dyes on titania-based photocatalysts by adding commercial GeO2 in aqueous suspension.
Chemical communications (Cambridge, England) (2010-06-25)
Jiaqiang Wang, Yuyang He, Jun Tao, Jiao He, Wenjun Zhang, Sijiang Niu, Zhiying Yan
ABSTRACT
Adding a small amount of commercial GeO(2) into aqueous suspension significantly enhanced the photocatalytic activity of titania-based photocatalysts for the degradation of dyes.
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Pricing and availability is not currently available.