Merck
  • Synthesis of N-, S-, and C-glycoside castanospermine analogues with selective neutral alpha-glucosidase inhibitory activity as antitumour agents.
Synthesis of N-, S-, and C-glycoside castanospermine analogues with selective neutral alpha-glucosidase inhibitory activity as antitumour agents.
Chemical communications (Cambridge, England) (2010-06-17)
Elena M Sánchez-Fernández, Rocío Rísquez-Cuadro, Maud Chasseraud, Ahmed Ahidouch, Carmen Ortiz Mellet, Halima Ouadid-Ahidouch, José M García Fernández
ABSTRACT
sp(2)-Iminosugar-type castanospermine analogues bearing an alpha-configured N-, S-, or C-linked pseudoanomeric group have been designed as selective inhibitors of the neutral alpha-glucosidases involved in N-glycoprotein processing; evaluation in breast cancer cell growth indicated a significant antiproliferative potential that was dependent on the nature of the pseudoanomeric group.
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%