Merck
  • Well-defined synthetic polymers with a protein-like gelation behavior in water.
Well-defined synthetic polymers with a protein-like gelation behavior in water.
Chemical communications (Cambridge, England) (2010-05-21)
Stefan Glatzel, Nezha Badi, Michael Päch, André Laschewsky, Jean-François Lutz
ABSTRACT
Homopolymers of N-acryloyl glycinamide were prepared by reversible addition-fragmentation chain transfer polymerization in water. The formed macromolecules exhibit strong polymer-polymer interactions in aqueous milieu and therefore form thermoreversible physical hydrogels in pure water, physiological buffer or cell medium.
Sigma-Aldrich
Glycinamide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Glycinamide hydrochloride, 98%