Merck
Cosmeceuticals: the evidence behind the retinoids.
Aesthetic surgery journal (2010-05-06)
Kajal Babamiri, Reza Nassab
ABSTRACT
A wide range of cosmeceutical products are available on the market currently, but evidence to support their use is often lacking in the literature. Specifically, there is a substantial amount of evidence supporting the efficacy of tretinoin in photoaging, but the evidence supporting retinoid-based cosmeceuticals remains sparse. The authors review the current data in the literature related to vitamin A-derived cosmeceutical products and conclude that cosmeceuticals containing retinaldehyde have been shown in large randomized, controlled trials to have the most beneficial effect on aging skin.
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g