Merck
  • Oxidation state, aggregation, and heterolytic dissociation of allyl indium reagents.
Oxidation state, aggregation, and heterolytic dissociation of allyl indium reagents.
Journal of the American Chemical Society (2010-04-10)
Konrad Koszinowski
ABSTRACT
Solutions of allyl indium reagents formed in the reactions of indium with allyl bromide and allyl iodide, respectively, in N,N-dimethylformamide, tetrahydrofuran, and water were analyzed by a combination of electrospray-ionization mass spectrometry, temperature-dependent (1)H NMR spectroscopy, and electrical conductivity measurements. Additional mass spectrometric experiments probed charge-tagged derivatives of the allyl indium reagents. The results obtained indicate the presence of allyl indium(+3) species, which undergo heterolytic dissociation to yield ions such as InR(2)(solv)(+) and InRX(3)(-) with R = allyl and X = Br and I. The extent of dissociation is greatest for N,N-dimethylformamide, whereas aggregation effects are more pronounced for the less polar tetrahydrofuran. The heterolytic dissociation of the allyl indium reagents supposedly enhances their reactivity by simultaneously providing highly Lewis acidic allyl indium cations and nucleophilic allyl indate anions.
Sigma-Aldrich
Allyl bromide, reagent grade, 97%, contains ≤1000 ppm propylene oxide as stabilizer