Merck
  • Synthesis of new beta-1-C-alkylated imino-L-iditols: A comparative study of their activity as beta-glucocerebrosidase inhibitors.
Synthesis of new beta-1-C-alkylated imino-L-iditols: A comparative study of their activity as beta-glucocerebrosidase inhibitors.
Bioorganic & medicinal chemistry (2010-03-17)
Wojciech Schönemann, Estelle Gallienne, Philippe Compain, Kyoko Ikeda, Naoki Asano, Olivier R Martin
ABSTRACT
A short synthesis of new beta-1-C-alkyl-1,5-dideoxy-1,5-imino-l-iditols by means of the diastereoselective addition of Grignard reagents onto a glucopyranosylamine is described. These compounds were evaluated as beta-glucocerebrosidase inhibitors and their activity was compared with that of related iminosugar derivatives in the d-gluco and d-xylo series. The results allowed us to conclude on the influence of the hydroxymethyl moiety and of the piperidine-ring conformation on the inhibitory activity.
Sigma-Aldrich
L-Iditol, ≥98% (GC)