Merck
  • Third-order nonlinear optical properties and structures of (E)-N-(4-nitrobenzylidene)-2,6-dimethylaniline and (E)-N-(4-nitrobenzylidene)-2,3-dimethylaniline.
Third-order nonlinear optical properties and structures of (E)-N-(4-nitrobenzylidene)-2,6-dimethylaniline and (E)-N-(4-nitrobenzylidene)-2,3-dimethylaniline.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-03-03)
Asli Karakas, Hüseyin Unver
ABSTRACT
(E)-N-(4-Nitrobenzylidene)-2,6-dimethylaniline (1) and (E)-N-(4-nitrobenzylidene)-2,3-dimethylaniline (2) have been synthesized. The crystal structures of both compounds have been defined by X-ray diffraction analysis. The maximum one-photon absorption (OPA) wavelengths recorded by quantum mechanical computations using a configuration interaction (CI) method are estimated in the UV region to be shorter than 450nm, showing good optical transparency to the visible light. To provide an insight into the microscopic third-order nonlinear optical (NLO) properties of the investigated molecules, both dispersion-free (static) and also frequency-dependent (dynamic) linear polarizabilities (alpha) and second hyperpolarizabilities (gamma) at lambda=825-1125nm and 1050-1600nm wavelength areas have been computed using time-dependent Hartree-Fock (TDHF) method. According to the ab initio calculation results, the title molecules exhibit second hyperpolarizabilities with non-zero values, implying microscopic third-order NLO behavior.
Sigma-Aldrich
2,6-Dimethylaniline, 99%
Supelco
2,6-Dimethylaniline, PESTANAL®, analytical standard