Merck
  • Redox-active ligands facilitate bimetallic O2 homolysis at five-coordinate oxorhenium(V) centers.
Redox-active ligands facilitate bimetallic O2 homolysis at five-coordinate oxorhenium(V) centers.
Journal of the American Chemical Society (2010-03-03)
Cameron A Lippert, Stephen A Arnstein, C David Sherrill, Jake D Soper
ABSTRACT
Five-coordinate oxorhenium(V) anions with redox-active catecholate and amidophenolate ligands are shown to effect clean bimetallic cleavage of O(2) to give dioxorhenium(VII) products. A structural homologue with redox-inert oxalate ligands does not react with O(2). Redox-active ligands lower the kinetic barrier to bimetallic O(2) homolysis at five-coordinate oxorhenium(V) by facilitating formation and stabilization of intermediate O(2) adducts. O(2) activation occurs by two sequential Re-O bond forming reactions, which generate mononuclear eta(1)-superoxo species, and then binuclear trans-mu-1,2-peroxo-bridged complexes. Formation of both Re-O bonds requires trapping of a triplet radical dioxygen species by a cis-[Re(V)(O)(cat)(2)](-) anion. In each reaction the dioxygen fragment is reduced by 1e(-), so generation of each new Re-O bond requires that an oxometal fragment is oxidized by 1e(-). Complexes containing a redox-active ligand access a lower energy reaction pathway for the 1e(-) Re-O bond forming reaction because the metal fragment can be oxidized without a change in formal rhenium oxidation state. It is also likely that redox-active ligands facilitate O(2) homolysis by lowering the barrier to the formally spin-forbidden reactions of triplet dioxygen with the closed shell oxorhenium(V) anions. By orthogonalizing 1e(-) and 2e(-) redox at oxorhenium(V), the redox-active ligand allows high-valent rhenium to utilize a mechanism for O(2) activation that is atypical of oxorhenium(V) but more typical for oxygenase enzymes and models based on 3d transition metal ions: O(2) cleavage occurs by a net 2e(-) process through a series of 1e(-) steps. The implications for design of new multielectron catalysts for oxygenase-type O(2) activation, as well as the microscopic reverse reaction, O-O bond formation from coupling of two M=O fragments for catalytic water oxidation, are discussed.
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%
Sigma-Aldrich
Ammonium perrhenate, ≥99%
Sigma-Aldrich
Ammonium perrhenate, 99.999% trace metals basis
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis