Merck
  • Efficacy of Ovsynch protocol with antiprolactin treatment for timed artificial insemination during non-breeding seasons in yaks (Poephagus grunniens L.).
Efficacy of Ovsynch protocol with antiprolactin treatment for timed artificial insemination during non-breeding seasons in yaks (Poephagus grunniens L.).
Reproduction in domestic animals = Zuchthygiene (2009-09-09)
M Sarkar, B K Dutta Borah, B S Prakash
ABSTRACT
An attempt was undertaken to investigate the efficacy of Ovsynch protocol for timed artificial insemination (TAI) with or without Norprolac (antiprolactin) treatment during non-breeding season (winter months) in yaks (n = 25). During non-breeding season, plasma prolactin profile has been reported high due to cold and nutritional stress. The Norprolac dose of i.m. administration was standardized for prolactin suppression. Three different doses viz. 2.5, 5.0 and 7.5 mg were attempted and the dose of 7.5 mg Norprolac i.m. per animal was found to be suitable for suppression of prolactin levels up to 30 h. Ovsynch treatment with Norprolac induced more number of oestrous symptoms per animal (4.8 vs 2.1), higher LH peak concentration (24.01 vs 16.16 ng/ml), longer duration of LH surge (6.8 vs 5.2 h) and higher conception rate (70 vs 30%) in Ovsynch plus Norprolac treated animals compared with animals treated with Ovsynch alone. Therefore, this study clearly indicates the opportunity for practical application of the Ovsynch plus Norprolac protocol for TAI in yaks during non-breeding seasons.
Product Description
Dinoprost trometamol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Prostaglandin F tris salt, synthetic, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Prostaglandin F tris salt, ≥99%, synthetic, powder
Pricing and availability is not currently available.